Assay Details
Target Gene Details
Entrez Gene ID: | 74694 |
Gene Name: | TBC1 domain family, member 30 |
Gene Aliases: |
4930505D03Rik |
Location: |
Chr.10:121263819-121311189 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d30 | NM_029057.1 | 12 | 2534 | NP_083333.1 |
| AK015700.1 | 6 | 1267 | ||
| AK044521.1 | 12 | 2341 | BAC31963.1 | |
| AK049989.1 | 12 | 2367 | ||
| AK173079.1 | 12 | 2297 | BAD32357.1 | |
| BC059081.1 | 4 | 1042 | AAH59081.1 | |
| BC079587.1 | 9 | 1843 | AAH79587.1 | |
| BC138285.1 | 12 | 2741 | ||
| BC138286.1 | 12 | 2741 |
More Information
Set Membership: |
|

Genomic Map