Assay Details
Target Gene Details
Entrez Gene ID: | 100504474 |
Gene Name: | small leucine-rich protein 1 |
Gene Aliases: |
2010003K15Rik |
Location: |
Chr.10:25527943-25536272 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smlr1 | NM_001195596.1 | 1 | 262 | NP_001182525.1 |
| AK008073.1 | 1 | 260 | ||
| AK080199.1 | 1 | 134 |

Genomic Map