Assay Details
Target Gene Details
Entrez Gene ID: | 15275 |
Gene Name: | hexokinase 1 |
Gene Aliases: |
BB404130, Hk-1, Hk1-s, dea, mHk1-s |
Location: |
Chr.10:62268855-62379920 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hk1 | NM_010438.3 | NP_034568.2 | ||
| EU430521.1 | ||||
| EU430522.1 | ||||
| EU430523.1 | ||||
| EU430524.1 | ||||
| EU430525.1 | ||||
| EU430526.1 | ||||
| EU430527.1 | ||||
| EU430528.1 | ||||
| EU430529.1 | ||||
| EU430530.1 | ||||
| EU430531.1 | ||||
| EU430532.1 | ||||
| EU430533.1 | ||||
| L16948.1 | AAB57760.1 | |||
| L16949.1 | AAB57759.1 | |||
| L16950.1 | AAA53036.1 |
More Information
Set Membership: |
|

Genomic Map