Assay Details
Target Gene Details
Entrez Gene ID: | 54140 |
Gene Name: | arginine vasopressin receptor 1A |
Gene Aliases: |
AVPR, Avpr1, V1a, V1aR |
Location: |
Chr.10:122448499-122453453 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Avpr1a | NM_016847.2 | 1 | 857 | NP_058543.2 |
| AK137203.1 | 1 | 785 | BAE23266.1 | |
| AK149578.1 | 1 | 800 | BAE28970.1 | |
| AK155952.1 | 1 | 790 | BAE33518.1 | |
| BC024149.1 | 1 | 855 | AAH24149.1 | |
| D49729.1 | 1 | 839 | BAA08566.1 | |
| D49730.1 | 1 | 864 | BAA08567.1 |
More Information
Set Membership: |
|

Genomic Map