Assay Details
Target Gene Details
Entrez Gene ID: | 56307 |
Gene Name: | methionine aminopeptidase 2 |
Gene Aliases: |
4930584B20Rik, A930035J23Rik, AI047573, AL024412, AU014659, Amp2, Mnpep, p67, p67eIF2 |
Location: |
Chr.10:93858489-93897090 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Metap2 | NM_019648.3 | 12 | 3440 | NP_062622.1 |
| AK053691.1 | 3 | 2821 | ||
| AK080746.1 | ||||
| AK165611.1 | 2 | 1618 | BAE38293.1 |
Target Gene Details
Entrez Gene ID: | 327799 |
Gene Name: | ubiquitin specific peptidase 44 |
Gene Aliases: |
E430004F17Rik |
Location: |
Chr.10:93819968-93860738 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp44 | AK041893.1 | 2 | 2466 |
More Information
Set Membership: |
|

Genomic Map