Assay Details
Target Gene Details
Entrez Gene ID: | 56307 |
Gene Name: | methionine aminopeptidase 2 |
Gene Aliases: |
4930584B20Rik, A930035J23Rik, AI047573, AL024412, AU014659, Amp2, Mnpep, p67, p67eIF2 |
Location: |
Chr.10:93858489-93897090 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Metap2 | NM_019648.3 | 2 | 353 | NP_062622.1 |
| AK016346.1 | ||||
| AK165301.1 | 2 | 354 |
More Information
Set Membership: |
|

Genomic Map