Assay Details
Target Gene Details
Entrez Gene ID: | 66125 |
Gene Name: | splicing factor 3b, subunit 5 |
Gene Aliases: |
10kDa, 1110005L13Rik, AU043053, Sf3b10 |
Location: |
Chr.10:13008450-13009183 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sf3b5 | NM_175102.4 | 1 | 611 | NP_780311.2 |
| AI849573.1 | 1 | 140 | ||
| AK003475.1 | 1 | 580 | BAC25037.1 | |
| AK009874.1 | 1 | 574 | BAC25275.1 | |
| AK010348.1 | 1 | 586 | BAC25292.1 | |
| BB652104.1 | 1 | 611 | ||
| BC006603.1 | 1 | 607 | AAH06603.1 |
More Information
Set Membership: |
|

Genomic Map