Assay Details
Target Gene Details
Entrez Gene ID: | 70312 |
Gene Name: | cactin, spliceosome C complex subunit |
Gene Aliases: |
2510012J08Rik |
Location: |
Chr.10:81321103-81326251 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cactin | NM_027381.2 | 9 | 2466 | NP_081657.1 |
| AK010963.1 | 6 | 1275 | BAB27295.1 | |
| AY934504.1 | 9 | 2372 | ||
| BC150986.1 | ||||
| BC151180.1 | ||||
| BY651766.1 | 1 | 161 |
More Information
Set Membership: |
|

Genomic Map