Assay Details
Target Gene Details
Entrez Gene ID: | 67307 |
Gene Name: | phenazine biosynthesis-like protein domain containing 2 |
Gene Aliases: |
3110049J23Rik, Mawbp |
Location: |
Chr.10:63024049-63058813 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pbld2 | NM_026085.2 | 10 | 1430 | NP_080361.1 |
| AK007784.1 | 9 | 1185 | ||
| AK014198.1 | 10 | 1431 | BAB29200.1 | |
| AK018692.1 | 9 | 1186 | BAB31348.1 | |
| BC061073.1 | 9 | 1185 | AAH61073.1 |

Genomic Map