Assay Details
Target Gene Details
Entrez Gene ID: | 74747 |
Gene Name: | DNA-damage-inducible transcript 4 |
Gene Aliases: |
5830413E08Rik, AA415483, REDD1, Rtp801, dig2 |
Location: |
Chr.10:59949675-59951770 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddit4 | NM_029083.2 | 3 | 790 | NP_083359.1 |
| AK017926.1 | 3 | 790 | BAB31006.1 | |
| AK081046.1 | 3 | 854 | BAC38121.1 | |
| AY260552.1 | 2 | 713 | AAP13851.1 | |
| BC131992.1 | 3 | 789 | ||
| BC132645.1 | 3 | 789 | ||
| BC145370.1 | 3 | 789 |

Genomic Map