Assay Details
Target Gene Details
Entrez Gene ID: | 56200 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 21 |
Gene Aliases: |
AI255159, AL022742, D10Ertd645e, D10Wsu42e |
Location: |
Chr.10:62580247-62602298 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx21 | NM_019553.2 | 2 | 394 | NP_062426.2 |
| AF159131.3 | 2 | 355 | AAD43959.3 | |
| AK010281.1 | 2 | 274 | BAB26817.2 | |
| AK012125.1 | BAB28047.2 | |||
| AK145578.1 | 2 | 378 | BAE26522.1 | |
| AK160095.1 | 2 | 368 | BAE35625.1 | |
| BC043655.1 | 2 | 366 | ||
| BC059237.1 | 2 | 354 | AAH59237.1 | |
| CF182162.1 | 2 | 411 |

Genomic Map