Assay Details
Target Gene Details
Entrez Gene ID: | 246086 |
Gene Name: | one cut domain, family member 3 |
Gene Aliases: |
OC-3, Oc3 |
Location: |
Chr.10:80494906-80517266 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Onecut3 | NM_139226.3 | 1 | 428 | NP_631972.2 |
| AY080897.1 | 1 | 380 | AAL86921.1 | |
| BC058700.1 | 1 | 428 | AAH58700.1 |
More Information
Set Membership: |
|

Genomic Map