Assay Details
Target Gene Details
Entrez Gene ID: | 56709 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily B, member 12 |
Gene Aliases: |
Dj10, mDj10 |
Location: |
Chr.10:59879569-59899296 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajb12 | NM_019965.2 | 9 | 1419 | NP_064349.2 |
| AK038149.1 | 8 | 1294 | ||
| AK046160.1 | ||||
| AK089427.1 | 9 | 1392 | ||
| AK138495.1 | 9 | 1349 | BAE23683.1 | |
| AK172274.1 | 9 | 1406 | BAE42919.1 | |
| AK172485.1 | 9 | 1381 | ||
| BC034162.1 | 9 | 1419 | AAH34162.1 |

Genomic Map