Assay Details
Target Gene Details
Entrez Gene ID: | 102115 |
Gene Name: | deoxyhypusine hydroxylase/monooxygenase |
Gene Aliases: |
1110033C18Rik, AI585913, AL022869, Hlrc1 |
Location: |
Chr.10:81384428-81388352 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dohh | NM_133964.2 | 2 | 313 | NP_598725.2 |
| AK004052.1 | 2 | 279 | BAC25064.1 | |
| AK154104.1 | 2 | 314 | ||
| AK202978.1 | 1 | 128 | ||
| BC002295.1 | 2 | 230 | AAH02295.1 | |
| BY131233.1 |
More Information
Set Membership: |
|

Genomic Map