Assay Details
Target Gene Details
Entrez Gene ID: | 67412 |
Gene Name: | SOGA family member 3 |
Gene Aliases: |
6330407J23Rik |
Location: |
Chr.10:29143274-29199630 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Soga3 | NM_026138.2 | 6 | 3071 | NP_080414.2 |
| AK012378.1 | 1 | 539 | BAB28200.1 | |
| AK018139.1 | 5 | 1601 | BAB31090.1 | |
| AK030682.1 | 6 | 2944 | ||
| AK163577.1 | 5 | 1574 | ||
| BC066079.1 | 6 | 3228 | AAH66079.1 |

Genomic Map