Assay Details
Target Gene Details
Entrez Gene ID: | 80904 |
Gene Name: | deltex 3 homolog (Drosophila) |
Gene Aliases: |
Deltex3, mg310 |
Location: |
Chr.10:127190378-127196412 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dtx3 | NM_030714.2 | 4 | 573 | NP_109639.1 |
| AB015425.1 | 3 | 555 | BAB18942.1 | |
| AK148444.1 | 4 | 592 | BAE28557.1 | |
| BC099687.1 | 4 | 573 | AAH99687.1 | |
| BC138304.1 | 3 | 452 | ||
| BC157899.1 | 3 | 454 |
More Information
Set Membership: |
|

Genomic Map