Assay Details
Target Gene Details
Entrez Gene ID: | 80904 |
Gene Name: | deltex 3 homolog (Drosophila) |
Gene Aliases: |
Deltex3, mg310 |
Location: |
Chr.10:127190378-127196412 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dtx3 | NM_030714.2 | 6 | 1616 | NP_109639.1 |
| AB015425.1 | 5 | 1598 | BAB18942.1 | |
| AK148444.1 | 5 | 1931 | BAE28557.1 | |
| AK205963.1 | 1 | 186 | ||
| AK212525.1 | 1 | 202 | ||
| AK213413.1 | 1 | 202 | ||
| AK217286.1 | 1 | 159 | ||
| BC099687.1 | 6 | 1616 | AAH99687.1 | |
| BC138304.1 | 5 | 1495 | ||
| BC157899.1 | 5 | 1497 |
More Information
Set Membership: |
|

Genomic Map