Assay Details
Target Gene Details
Entrez Gene ID: | 18451 |
Gene Name: | procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha 1 polypeptide |
Gene Aliases: |
AL022634, P4ha |
Location: |
Chr.10:59323246-59373304 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P4ha1 | NM_011030.2 | 15 | 2889 | NP_035160.1 |
| AK030574.1 | 15 | 2818 | ||
| AK045008.1 | 15 | 2818 | BAC32183.1 | |
| AK149127.1 | 15 | 2781 | BAE28745.1 | |
| AK160798.1 | 15 | 2818 | BAE36020.1 | |
| BC009654.1 | 15 | 2812 | AAH09654.1 |
More Information
Set Membership: |
|

Genomic Map