Assay Details
Target Gene Details
Entrez Gene ID: | 114774 |
Gene Name: | PRKC, apoptosis, WT1, regulator |
Gene Aliases: |
2310001G03Rik, PAR4, Par-4 |
Location: |
Chr.10:108332189-108414391 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pawr | NM_054056.2 | 2 | 584 | NP_473397.1 |
| AF377871.1 | AAK55414.1 | |||
| AW123400.1 | 1 | 187 | ||
| DQ363525.1 | 2 | 584 | ABC96645.1 | |
| DQ449073.1 | 1 | 395 | ||
| DQ449074.1 | 1 | 395 |

Genomic Map