Assay Details
Target Gene Details
Entrez Gene ID: | 76877 |
Gene Name: | RAB36, member RAS oncogene family |
Gene Aliases: |
6330593L16Rik, AW047545 |
Location: |
Chr.10:75036895-75054748 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab36 | NM_029781.3 | 11 | 1253 | NP_084057.1 |
| AK018269.1 | 11 | 1271 | BAB31140.1 | |
| AK038445.1 | 11 | 1254 | BAC30001.1 | |
| AK043946.1 | 11 | 1277 | BAE20645.1 | |
| AK046113.1 | 11 | 1062 | BAE20653.1 | |
| AK142025.1 | 10 | 1625 | BAE24917.1 |

Genomic Map