Assay Details
Target Gene Details
Entrez Gene ID: | 12955 |
Gene Name: | crystallin, alpha B |
Gene Aliases: |
Crya-2, Crya2, HspB5, P23 |
Location: |
Chr.9:50751325-50756636 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cryab | NM_001289782.1 | NP_001276711.1 | ||
| BC094033.1 | AAH94033.1 |
Target Gene Details
Entrez Gene ID: | 69253 |
Gene Name: | heat shock protein 2 |
Gene Aliases: |
27kDa, 2810021G24Rik, HSP27, MKBP |
Location: |
Chr.9:50751072-50752354 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hspb2 | NM_001164708.1 | 2 | 342 | NP_001158180.1 |
| NM_024441.3 | 2 | 624 | NP_077761.3 | |
| AK012780.1 | 2 | 343 | BAB28465.1 | |
| BC064051.1 | 2 | 563 | AAH64051.1 |

Genomic Map