Assay Details
Target Gene Details
Entrez Gene ID: | 52679 |
Gene Name: | E2F transcription factor 7 |
Gene Aliases: |
A630014C11Rik, D10Ertd739e |
Location: |
Chr.10:110745427-110787384 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E2f7 | NM_178609.4 | 12 | 2689 | NP_848724.2 |
| AK031078.1 | 12 | 2687 | BAC27243.1 | |
| AK045040.1 | 12 | 2654 | BAC32193.1 | |
| AK143686.1 | 12 | 2662 | BAE25497.1 | |
| AY463359.1 | 12 | 2686 | AAR26542.1 | |
| BC145429.1 | 12 | 2642 | ||
| BC150772.1 | 12 | 2642 |
More Information
Set Membership: |
|

Genomic Map