Assay Details
Target Gene Details
Entrez Gene ID: | 68114 |
Gene Name: | melanoma associated antigen (mutated) 1 |
Gene Aliases: |
2610019J09, 9430059D04Rik, UBE-1C2 |
Location: |
Chr.10:80226434-80245144 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mum1 | NM_023431.5 | 12 | 1918 | NP_075920.4 |
| AK011465.1 | 3 | 422 | BAB27636.1 | |
| AK020470.1 | 2 | 316 | BAB32113.1 | |
| AK078792.1 | 9 | 1625 | BAC37397.1 | |
| AK147490.1 | 2 | 5073 | ||
| AK154610.1 | 11 | 2305 | ||
| AK158750.1 | 12 | 1918 | BAE34640.1 | |
| AK159972.1 | 11 | 2820 | ||
| BC023031.1 | 7 | 676 | AAH23031.1 | |
| BC069883.1 | 5 | 554 | AAH69883.1 | |
| BC075617.1 | 11 | 1807 | AAH75617.1 |
More Information
Set Membership: |
|

Genomic Map