Assay Details
Target Gene Details
Entrez Gene ID: | 216363 |
Gene Name: | RAB3A interacting protein |
Gene Aliases: |
B230311A06, Gtpat12, Rabin3 |
Location: |
Chr.10:116905780-116950448 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab3ip | NM_001003950.2 | 3 | 576 | NP_001003950.1 |
| AK152122.1 | 3 | 628 | ||
| AK178592.1 | 1 | 136 | ||
| AK203553.1 | 1 | 136 | ||
| BC080289.1 | 3 | 576 | AAH80289.1 | |
| S79539.1 | AAB35170.1 |
More Information
Set Membership: |
|

Genomic Map