Assay Details
Target Gene Details
Entrez Gene ID: | 270160 |
Gene Name: | RAB39, member RAS oncogene family |
Gene Aliases: |
C230094F14Rik, Rab39a |
Location: |
Chr.9:53684110-53706232 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab39 | NM_175562.3 | 2 | 785 | NP_780771.1 |
| AB232635.1 | BAF02897.1 | |||
| AK041076.1 | 1 | 362 | BAC30810.1 | |
| AK049038.1 | 2 | 785 | BAC33522.1 | |
| AK053487.1 | 2 | 782 | BAC35399.1 | |
| BC049787.1 | 2 | 765 | AAH49787.1 |

Genomic Map