Assay Details
Target Gene Details
Entrez Gene ID: | 14385 |
Gene Name: | solute carrier family 37 (glucose-6-phosphate transporter), member 4 |
Gene Aliases: |
G6PT, G6pt1, GSD-1b, mG6PT |
Location: |
Chr.9:44396857-44402968 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc37a4 | NM_001293630.1 | 9 | 1799 | NP_001280559.1 |
| NM_001293631.1 | 9 | 1512 | NP_001280560.1 | |
| NM_008063.2 | 8 | 1539 | NP_032089.2 | |
| AF080469.1 | 8 | 1360 | AAC79840.1 | |
| AK003620.1 | 8 | 1362 | BAB22894.1 | |
| AK147849.1 | 9 | 1469 | ||
| AK155339.1 | 9 | 1796 | BAE33203.1 | |
| BC075655.1 | 8 | 1539 | AAH75655.1 |
More Information
Set Membership: |
|

Genomic Map