Assay Details
Target Gene Details
Entrez Gene ID: | 69718 |
Gene Name: | inositol polyphosphate multikinase |
Gene Aliases: |
2410017C19Rik, AA408208, Impk |
Location: |
Chr.10:71347765-71385885 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ipmk | NM_027184.1 | 6 | 3073 | NP_081460.1 |
| AK034274.1 | 1 | 1176 | ||
| AK036084.1 | 6 | 2841 | BAC29300.1 | |
| BC052463.1 | 6 | 3073 | AAH52463.1 |
More Information
Set Membership: |
|

Genomic Map