Assay Details
Target Gene Details
Entrez Gene ID: | 70784 |
Gene Name: | RAS-like, family 12 |
Gene Aliases: |
4631404I11Rik |
Location: |
Chr.9:65398485-65415060 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rasl12 | NM_001162923.1 | 5 | 1707 | NP_001156395.1 |
| AK048176.1 | 5 | 1661 |
Target Gene Details
Entrez Gene ID: | 330962 |
Gene Name: | solute carrier family 51, beta subunit |
Gene Aliases: |
Ostb |
Location: |
Chr.9:65412753-65422773 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc51b | NM_178933.2 | NP_849264.1 | ||
| AK002463.1 | BAE20415.1 | |||
| AK004188.1 | BAE20418.1 | |||
| AY279396.1 | AAP23994.1 | |||
| BC115606.1 | ||||
| BC115607.1 |
More Information
Set Membership: |
|

Genomic Map