Assay Details
Target Gene Details
Entrez Gene ID: | 268345 |
Gene Name: | potassium voltage gated channel, Shaw-related subfamily, member 2 |
Gene Aliases: |
AW047325, B230117I07, KShIIIA, Kv3.2 |
Location: |
Chr.10:112270459-112466304 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnc2 | NM_001025581.1 | 2 | 165 | NP_001020752.1 |
| AK045425.1 | 2 | 168 | ||
| BC116289.1 | 2 | 167 | ||
| BC116290.1 | 2 | 166 |
More Information
Set Membership: |
|

Genomic Map