Assay Details
Target Gene Details
Entrez Gene ID: | 17938 |
Gene Name: | nascent polypeptide-associated complex alpha polypeptide |
Gene Aliases: |
AL022831, AL024382, Gm1878, mKIAA0363, skNAC |
Location: |
Chr.10:128035346-128048637 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Naca | NM_001113199.1 | 1 | 169 | NP_001106670.1 |
| NM_001282976.1 | 1 | 169 | NP_001269905.1 | |
| NM_013608.3 | 1 | 169 | NP_038636.2 | |
| BU962928.1 | 1 | 143 |
More Information
Set Membership: |
|

Genomic Map