Assay Details
Target Gene Details
Entrez Gene ID: | 19682 |
Gene Name: | retinol dehydrogenase 5 |
Gene Aliases: |
9-cis, AI987873, RDH4, cRDH |
Location: |
Chr.10:128913588-128919297 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rdh5 | NM_134006.4 | 5 | 1092 | NP_598767.1 |
| AF013288.1 | 5 | 963 | AAC25951.1 | |
| AF033195.1 | 5 | 1030 | AAC00491.1 | |
| AF033196.2 | 5 | 1058 | AAC00492.1 | |
| AI838677.1 | 1 | 184 | ||
| AK135139.1 | 5 | 1093 | BAE22435.1 | |
| BC021372.1 | 5 | 1005 | AAH21372.1 |
More Information
Set Membership: |
|

Genomic Map