Assay Details
Target Gene Details
Entrez Gene ID: | 11630 |
Gene Name: | absent in melanoma 1 |
Gene Aliases: |
AI462491, AI463325 |
Location: |
Chr.10:43950307-44148924 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aim1 | NM_172393.2 | 20 | 6867 | NP_765981.2 |
| AF397913.1 | 14 | 2575 | ||
| AK041082.1 | 19 | 3484 | ||
| AK088102.1 | 19 | 3539 | BAC40145.1 | |
| BC031829.1 | 7 | 1880 | AAH31829.1 | |
| BC035551.1 | 19 | 4019 | AAH35551.1 |

Genomic Map