Assay Details
Target Gene Details
Entrez Gene ID: | 67723 |
Gene Name: | centrosomal protein 83, opposite strand |
Gene Aliases: |
4932415G12Rik, Ccdc41os1 |
Location: |
Chr.10:94673493-94688613 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cep83os | NR_015524.1 | |||
| AK015136.1 | ||||
| AK016525.1 | ||||
| AK085032.1 | ||||
| AK144701.1 | ||||
| AK154718.1 | ||||
| AK155868.1 |
More Information
Set Membership: |
|

Genomic Map