Assay Details
Target Gene Details
Entrez Gene ID: | 114229 |
Gene Name: | KISS1 receptor |
Gene Aliases: |
Gpr54, KiSS-1 |
Location: |
Chr.10:79916971-79922282 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kiss1r | NM_053244.5 | 1 | 975 | NP_444474.1 |
| AK039628.1 | 1 | 976 | BAC30404.1 | |
| AK139303.1 | 1 | 976 |
More Information
Set Membership: |
|

Genomic Map