Assay Details
Target Gene Details
Entrez Gene ID: | 544710 |
Gene Name: | keratin associated protein 10-10 |
Gene Aliases: |
EG544710 |
Location: |
Chr.10:77835947-77837251 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtap10-10 | NM_001024709.3 | 1 | 858 | NP_001019880.2 |
| AK079449.1 | 1 | 859 | BAC37648.1 |
Target Gene Details
Entrez Gene ID: | 252974 |
Gene Name: | thrombospondin type laminin G domain and EAR repeats |
Gene Aliases: |
C330046G03Rik, ORF65, TSP-EAR, Tnep1 |
Location: |
Chr.10:77686030-77886994 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tspear | NM_001287074.1 | NP_001274003.1 | ||
| AJ487520.1 | CAD31788.1 | |||
| JQ815565.1 |
More Information
Set Membership: |
|

Genomic Map