Assay Details
Target Gene Details
Entrez Gene ID: | 16694 |
Gene Name: | keratin associated protein 12-1 |
Gene Aliases: |
D10Jhu14e |
Location: |
Chr.10:77720586-77721256 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtap12-1 | NM_010670.1 | 1 | 441 | NP_034800.1 |
| BC107319.1 | 1 | 428 | AAI07320.1 | |
| BC107320.1 | 1 | 428 | AAI07321.1 |
Target Gene Details
Entrez Gene ID: | 252974 |
Gene Name: | thrombospondin type laminin G domain and EAR repeats |
Gene Aliases: |
C330046G03Rik, ORF65, TSP-EAR, Tnep1 |
Location: |
Chr.10:77686030-77886994 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tspear | NM_001287074.1 | NP_001274003.1 | ||
| JQ815565.1 |
More Information
Set Membership: |
|

Genomic Map