Assay Details
Target Gene Details
Entrez Gene ID: | 211329 |
Gene Name: | nuclear receptor coactivator 7 |
Gene Aliases: |
9030406N13Rik |
Location: |
Chr.10:30645582-30805977 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncoa7 | NM_001111267.2 | 6 | 1027 | NP_001104737.1 |
| NM_172495.5 | 16 | 3378 | NP_766083.3 | |
| AK033503.1 | 6 | 941 | BAC28325.1 | |
| AK042369.1 | 5 | 1312 | BAC31241.1 | |
| AK128935.1 | 15 | 3201 | BAC87660.1 | |
| AK165098.1 | 4 | 1715 | ||
| AK171679.1 | 5 | 871 | BAE42607.1 | |
| BC049997.1 | 6 | 977 | AAH49997.1 | |
| BC058572.1 | 6 | 949 | AAH58572.1 | |
| BC076623.1 | 14 | 2984 | AAH76623.1 | |
| EF602028.1 | 6 | 1027 |

Genomic Map