Assay Details
Target Gene Details
Entrez Gene ID: | 66932 |
Gene Name: | REX1, RNA exonuclease 1 homolog (S. cerevisiae) |
Gene Aliases: |
1700021P10Rik, 2610511M11Rik, AW050347, EloA-BP1, Tceb3bp1 |
Location: |
Chr.10:80540926-80563381 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rexo1 | NM_025852.3 | 1 | 102 | NP_080128.2 |
| AK012127.1 | 1 | 84 | BAC25356.1 | |
| AK087016.1 | 1 | 85 | BAC39784.1 | |
| AK144769.1 | 1 | 89 | BAE26057.1 | |
| AK173108.1 | 1 | 67 | BAD32386.1 | |
| BC025511.1 | 1 | 73 | ||
| BC039623.1 | 1 | 53 | AAH39623.1 | |
| BC052424.1 | 1 | 84 | AAH52424.1 | |
| BC069964.1 | 1 | 54 | AAH69964.1 | |
| BY234048.1 | 1 | 102 |
More Information
Set Membership: |
|

Genomic Map