Assay Details
Target Gene Details
Entrez Gene ID: | 19659 |
Gene Name: | retinol binding protein 1, cellular |
Gene Aliases: |
CRBPI, Crbp, Rbp-1 |
Location: |
Chr.9:98422961-98446551 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbp1 | NM_011254.5 | 1 | 692 | NP_035384.1 |
| X60367.1 | 1 | 692 | CAA42919.1 |

Genomic Map