Assay Details
Target Gene Details
Entrez Gene ID: | 106947 |
Gene Name: | solute carrier family 39 (zinc transporter), member 3 |
Gene Aliases: |
AI845814, Zip3 |
Location: |
Chr.10:81028540-81037421 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc39a3 | NM_134135.1 | 2 | 1356 | NP_598896.1 |
| AK087519.1 | 4 | 1513 | ||
| AK194186.1 | 1 | 169 | ||
| BC005502.1 | 2 | 1356 | AAH05502.1 |
More Information
Set Membership: |
|

Genomic Map