Assay Details
Target Gene Details
Entrez Gene ID: | 72480 |
Gene Name: | TSPY-like 4 |
Gene Aliases: |
2610102M01Rik, B230210I21Rik, D10Bwg0791e |
Location: |
Chr.10:34297421-34301320 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tspyl4 | NM_030203.2 | 1 | 645 | NP_084479.1 |
| AK020999.1 | ||||
| AK030740.1 | 1 | 590 | BAC27108.1 | |
| AK034070.1 | 1 | 612 | BAC28571.1 | |
| AK034414.1 | 1 | 586 | BAC28701.1 | |
| AK138536.1 | 1 | 751 | ||
| AK158197.1 | 1 | 613 | ||
| BC017540.1 | 1 | 594 | AAH17540.1 | |
| BC030922.1 | 1 | 120 | AAH30922.1 | |
| BC034656.1 | 1 | 593 | AAH34656.1 |
More Information
Set Membership: |
|

Genomic Map