Assay Details
Target Gene Details
Entrez Gene ID: | 12534 |
Gene Name: | cyclin-dependent kinase 1 |
Gene Aliases: |
Cdc2, Cdc2a, p34<CDC2> |
Location: |
Chr.10:69336635-69352912 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk1 | NM_007659.3 | 8 | 1878 | NP_031685.2 |
| AK030231.1 | 8 | 1879 | BAC26856.1 |
More Information
Set Membership: |
|

Genomic Map