Assay Details
Target Gene Details
Entrez Gene ID: | 216169 |
Gene Name: | abhydrolase domain containing 17A |
Gene Aliases: |
1700013O15Rik, BC005632, D10Bwg1364e, Fam108a |
Location: |
Chr.10:80583649-80590341 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abhd17a | NM_145421.2 | 2 | 509 | NP_663396.1 |
| AK170785.1 | 2 | 493 | ||
| AK220316.1 | 1 | 3034 | BAD90391.1 | |
| BC005632.1 | 2 | 484 | AAH05632.1 | |
| BC082997.1 | 2 | 473 | AAH82997.1 |
More Information
Set Membership: |
|

Genomic Map