Assay Details
Target Gene Details
Entrez Gene ID: | 237397 |
Gene Name: | C2 calcium-dependent domain containing 4C |
Gene Aliases: |
4932409I22Rik, Fam148c, Gm238 |
Location: |
Chr.10:79606854-79614025 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C2cd4c | NM_001168624.1 | 2 | 3384 | NP_001162095.1 |
| NM_198614.3 | 2 | 3510 | NP_941016.2 | |
| BC089008.1 | 2 | 3492 | AAH89008.1 |

Genomic Map