Assay Details
Target Gene Details
Entrez Gene ID: | 66932 |
Gene Name: | REX1, RNA exonuclease 1 homolog (S. cerevisiae) |
Gene Aliases: |
1700021P10Rik, 2610511M11Rik, AW050347, EloA-BP1, Tceb3bp1 |
Location: |
Chr.10:80540926-80563381 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rexo1 | NM_025852.3 | 2 | 1168 | NP_080128.2 |
| AK012127.1 | 2 | 1149 | BAC25356.1 | |
| AK087016.1 | 2 | 1153 | BAC39784.1 | |
| AK144769.1 | 2 | 1155 | BAE26057.1 | |
| AK173108.1 | 2 | 1133 | BAD32386.1 | |
| BC025511.1 | 2 | 1139 | ||
| BC039623.1 | 2 | 1119 | AAH39623.1 | |
| BC052424.1 | 2 | 1150 | AAH52424.1 | |
| BC069964.1 | 2 | 1120 | AAH69964.1 |
More Information
Set Membership: |
|

Genomic Map