Assay Details
Target Gene Details
Entrez Gene ID: | 629557 |
Gene Name: | glyceraldehyde-3-phosphate dehydrogenase pseudogene |
Gene Aliases: |
EG629557 |
Location: |
Chr.9:52002061-52048705 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm6981 | NR_023357.1 | |||
| AK132984.1 |
Target Gene Details
Entrez Gene ID: | 19684 |
Gene Name: | radixin |
Gene Aliases: |
AA516625 |
Location: |
Chr.9:52047150-52088738 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rdx | NM_001104616.1 | NP_001098086.1 | ||
| NM_001104617.1 | 1 | 407 | NP_001098087.1 | |
| NM_009041.3 | 1 | 407 | NP_033067.2 | |
| AK016711.1 | 1 | 78 | ||
| AK088292.1 | 1 | 114 | BAC40263.2 | |
| AK146423.1 | 1 | 65 | BAE27157.1 | |
| AK156370.1 | 1 | 93 | BAE33689.1 | |
| AK162204.1 | BAE36790.1 | |||
| AK168449.1 | 1 | 78 | BAE40352.1 | |
| BC026786.1 | 1 | 80 | ||
| BC053417.1 | 1 | 81 | AAH53417.1 | |
| BY739389.1 | 1 | 407 | ||
| DV662898.1 | ||||
| X60672.1 | CAA43087.1 |
More Information
Set Membership: |
|

Genomic Map