Assay Details
Target Gene Details
Entrez Gene ID: | 107449 |
Gene Name: | unc-5 homolog B (C. elegans) |
Gene Aliases: |
6330415E02Rik, A630020F16, D10Bwg0792e, Unc5h2 |
Location: |
Chr.10:60762594-60831604 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Unc5b | NM_029770.2 | 10 | 1907 | NP_084046.2 |
| AJ487853.1 | 10 | 1810 | CAD32251.1 | |
| AK018177.1 | 10 | 1907 | BAB31108.1 | |
| AK147603.1 | 9 | 1769 | BAE28019.1 | |
| AK154271.1 | 10 | 1907 | BAE32479.1 | |
| AK154287.1 | 10 | 1910 | ||
| AK158284.1 | 10 | 1907 | BAE34445.1 | |
| BC048162.1 | 9 | 1681 | AAH48162.1 | |
| BC057560.1 | 9 | 1681 | AAH57560.1 |
More Information
Set Membership: |
|

Genomic Map