Assay Details
Target Gene Details
Entrez Gene ID: | 270711 |
Gene Name: | family with sequence similarity 26, member D |
Gene Aliases: |
4732454E20Rik, AI586164, Gm643 |
Location: |
Chr.10:34038784-34044313 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam26d | NM_001081165.1 | 2 | 3194 | NP_001074634.1 |
| AK028763.1 | 2 | 3188 | BAC26107.1 |
Target Gene Details
Entrez Gene ID: | 692132 |
Gene Name: | trafficking protein particle complex 3 like |
Gene Aliases: |
Bet3l, ENSMUSG00000071340 |
Location: |
Chr.10:34037597-34109815 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trappc3l | NM_001162937.1 | NP_001156409.1 | ||
| BC099502.1 | AAH99502.1 |
More Information
Set Membership: |
|

Genomic Map