Assay Details
Target Gene Details
Entrez Gene ID: | 103511 |
Gene Name: | family with sequence similarity 26, member E |
Gene Aliases: |
BB146404 |
Location: |
Chr.10:34091352-34096519 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam26e | NM_178908.3 | 1 | 412 | NP_849239.2 |
| AK078850.1 | 1 | 380 | BAC37420.1 | |
| AK085925.1 | 1 | 412 | BAC39570.1 | |
| AK161070.1 | 1 | 344 | BAE36184.1 | |
| BC025893.1 | 1 | 387 | AAH25893.1 |
Target Gene Details
Entrez Gene ID: | 692132 |
Gene Name: | trafficking protein particle complex 3 like |
Gene Aliases: |
Bet3l, ENSMUSG00000071340 |
Location: |
Chr.10:34037597-34109815 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trappc3l | NM_001162937.1 | NP_001156409.1 | ||
| BC099502.1 | AAH99502.1 |
More Information
Set Membership: |
|

Genomic Map