Assay Details
Target Gene Details
Entrez Gene ID: | 215900 |
Gene Name: | family with sequence similarity 26, member F |
Gene Aliases: |
A630077B13Rik |
Location: |
Chr.10:34126067-34127972 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam26f | NM_175449.4 | 2 | 663 | NP_780658.2 |
| AK042270.1 | 2 | 663 | BAC31209.1 | |
| AK080048.1 | 2 | 298 | BAC37815.1 | |
| AK153519.1 | 2 | 661 | ||
| BC095946.1 | 2 | 677 | AAH95946.1 |

Genomic Map